Order Entry
Netherlands
ContactUsLinkComponent
 
 
Ethyl malonyl chloride ≥97.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
Ethyl malonyl chloride ≥97.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Yellow to Green clear liquid
  • Purity (GC):
    min. 97.0 %
  • Purity (Argentmetric Titration):
    min. 97.0 %

 

Frequently Bought Together