
Decyltrimethylammonium bromide ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2082-84-0
:
Formula:C₁₃H₃₀BrN
MDL Number:MFCD00041973
Melting Pt:242°C
:280.30 g/mol
EINECS:218-219-7
:GHS07
Specification Test Results
- Appearance:White powder to crystal
- Purity (Nonaqueous Titration):min. 99.0 %
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together