

2,2-Dimethylpropane-1,3-diamine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:7328-91-8
:
Formula:C₅H₁₄N₂
MDL Number:MFCD00009801
:102.1800 g/mol
Boiling Pt:65°C
Flash Pt:47°C
EINECS:230-819-0
UN:2921
ADR:8,II
:GHS02
:GHS05
Specification Test Results
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Freezing point:25.0 to 28.0 deg-C
Specification Test Results
Frequently Bought Together