Order Entry
Netherlands
ContactUsLinkComponent
2-Isobutyl-3-methylpyrazine ≥98.0% (by GC)
Irritant
2-Isobutyl-3-methylpyrazine ≥98.0% (by GC)
Catalog # TCIAI0337-5ML
Supplier:  TCI
CAS Number:  
undefined
Irritant
2-Isobutyl-3-methylpyrazine ≥98.0% (by GC)
Catalog # TCIAI0337-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Orange to Green clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
Boiling Pt:92°C
Flash Pt:82°C
 :150.2209 g/mol
EINECS:237-693-6
 :GHS07
CAS Number:13925-06-9
Formula:C₉H₁₄N₂
MDL Number:MFCD00059770