
Diphenylboric acid-2-aminoethyl ester (2-APB) ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:524-95-8
:
Formula:C₁₄H₁₆BNO
MDL Number:MFCD00014823
:225.1 g/mol
EINECS:208-366-5
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:189.0 to 194.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together