
Naphthalene-1,4-dicarbonitrile ≥98.0% (by GC)
Catalog # TCIAD2758-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:209.0 to 213.0 deg-C
- Solubility in hot Toluene:very faint turbidity
Specification Test Results
About this item
:
:178.19 g/mol
EINECS:221-197-1
UN:3439
ADR:6.1,III
:GHS06
CAS Number:3029-30-9
Formula:C₁₂H₆N₂
MDL Number:MFCD03093626