
1,1-Diphenylhydrazine hydrochloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD0895-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Solubility in Methanol:within almost transparency
Specification Test Results
About this item
:
:220.70 g/mol
Merck Index:14,03325
EINECS:208-481-0
:GHS07
CAS Number:530-47-2
Formula:C₁₂H₁₃ClN₂
MDL Number:MFCD00012950