
1,1-Diphenylhydrazine ≥97.0% (by GC, titration analysis)
Catalog # TCIAD1161-5ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:White or Colorles to Yellow to Orange powder to lump to clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
About this item
:
Merck Index:14,03325
EINECS:208-483-1
:GHS07
CAS Number:530-50-7
Formula:C₁₂H₁₂N₂
MDL Number:MFCD00044505