
(2-Hydroxybenzyl)triphenylphosphonium bromide ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAH1240-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
About this item
:
:449.34 g/mol
Melting Pt:243°C
EINECS:274-574-8
:GHS07
CAS Number:70340-04-4
Formula:C₂₅H₂₂BrOP
MDL Number:MFCD00011902