
4-(Methylamino)pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1121-58-0
:
Formula:C₆H₈N₂
MDL Number:MFCD00152625
:108.14 g/mol
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:125.0 to 129.0 deg-C
Specification Test Results
Frequently Bought Together