
Isopropyltriphenylphosphonium iodide ≥98.0% (by HPLC, titration analysis)
: TCI
:24470-78-8
:
:C₂₁H₂₂IP
:MFCD00031595
:432.29 g/mol
:246-281-5
:GHS07
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:196.0 to 200.0 deg-C
- Solubility in Methanol:very faint turbidity
Frequently Bought Together