Order Entry
Netherlands
ContactUsLinkComponent
1,3-Diisopropylimidazolium tetrafluoroborate ≥96.0% (by total nitrogen basis)
Corrosive
Irritant
1,3-Diisopropylimidazolium tetrafluoroborate ≥96.0% (by total nitrogen basis)
Catalog # TCIAD3623-1G
Supplier:  TCI
CAS Number:  
Corrosive
Irritant
1,3-Diisopropylimidazolium tetrafluoroborate ≥96.0% (by total nitrogen basis)
Catalog # TCIAD3623-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (HPLC):
    min. 99.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    85.0 to 90.0 deg-C

Specification Test Results

About this item

 :
ADR:8,III
 :240.0493 g/mol
UN:1759
 :GHS05
 :GHS07
CAS Number:286014-34-4
Formula:C₉H₁₇BF₄N₂
MDL Number:MFCD08276804