Order Entry
Netherlands
ContactUsLinkComponent
Palmitic acid hydrazide ≥98.0% (by titrimetric analysis)
Irritant
Palmitic acid hydrazide ≥98.0% (by titrimetric analysis)
Catalog # TCIAP0004-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Palmitic acid hydrazide ≥98.0% (by titrimetric analysis)
Catalog # TCIAP0004-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Solubility in hot EtOH:
    almost transparency
  • Melting point:
    111.0 to 115.0 deg-C

Specification Test Results

About this item

 :
 :270.46 g/mol
 :GHS07
CAS Number:2619-88-7
Formula:C₁₆H₃₄N₂O
MDL Number:MFCD00066357