1,5-Naphthyridine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:254-79-5
:
Formula:C₈H₆N₂
MDL Number:MFCD00039727
:130.15 g/mol
Boiling Pt:152°C
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:73.0 to 78.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together