Order Entry
Netherlands
ContactUsLinkComponent
 
 
Allyltriphenylphosphonium bromide ≥98.0% (by titrimetric analysis)
  TCIAA1007-25G
 :  TCI
 :  
undefined
 
Allyltriphenylphosphonium bromide ≥98.0% (by titrimetric analysis)
  TCIAA1007-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

 

 :
 :383.261 g/mol
 :216-332-6
 :GHS07
 :1560-54-9
 :C₂₁H₂₀BrP
 :MFCD00011808