α-Methylcinnamaldehyde ≥95.0% (by GC)
Supplier: TCI
About this item
Synonyms: α-Methyl-cinnamaldehyde(predominantly trans)
- CAS Number:101-39-3
- Formula:C₁₀H₁₀O
- MDL Number:MFCD00006976
- Molecular Weight:146.19 g/mol
- Boiling Pt:132°C
- Flash Pt:124°C
- EINECS:202-938-8
2 Options Available Below
Product Details & Documents
Synonyms: α-Methyl-cinnamaldehyde(predominantly trans)
- CAS Number:101-39-3
- Formula:C₁₀H₁₀O
- MDL Number:MFCD00006976
- Molecular Weight:146.19 g/mol
- Boiling Pt:132°C
- Flash Pt:124°C
- EINECS:202-938-8
Specifications for Product Family
Specification Test Results
- Appearance:Light yellow to Yellow to Orange clear liquid
- Purity (GC):min. 95.0 %



