Order Entry
Netherlands
ContactUsLinkComponent
Methyl hydrogen glutarate ≥97.0% (by GC, titration analysis)
Methyl hydrogen glutarate ≥97.0% (by GC, titration analysis)
  TCIAG0258-25G
 :  TCI
 :  
undefined
Methyl hydrogen glutarate ≥97.0% (by GC, titration analysis)
  TCIAG0258-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization titration):
    min. 97.0 %

 

 

 :
 :146.1412 g/mol
 :158°C
 :216-116-1
 :1501-27-5
 :C₆H₁₀O₄
 :MFCD00004409