
(-)-Di-p-toluoyl-L-tartaric acid ≥98.0% (by HPLC, titration analysis)
: TCI
:32634-66-5
:
:C₂₀H₁₈O₈
:MFCD00064440
:386.36 g/mol
:251-131-7
:GHS07
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Optical purity (LC):min. 98.0 ee%
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Methanol:almost transparency
Frequently Bought Together