Order Entry
Netherlands
ContactUsLinkComponent
 
 
1,5-Dimethoxypentane ≥97.0% (by GC)
  TCIAD2857-5G
 :  TCI
 :  
undefined
 
1,5-Dimethoxypentane ≥97.0% (by GC)
  TCIAD2857-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %
  • Specific gravity (20/20):
    0.8570 to 0.8670

 

 

 :
 :132.200 g/mol
 :1993
 :3,III
 :GHS02
 :111-89-7
 :C₇H₁₆O₂
 :MFCD03613608