Order Entry
Netherlands
ContactUsLinkComponent
4,6-Dihydroxy-2-methylpyrimidine ≥96.0% (by titrimetric analysis)
Irritant
4,6-Dihydroxy-2-methylpyrimidine ≥96.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Irritant
4,6-Dihydroxy-2-methylpyrimidine ≥96.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (Neutralization titration):
    min. 96.0 %
  • Solubility in 1mol/L NaOH:
    almost transparency

Specification Test Results

Frequently Bought Together