

1,2-Dichloro-1,1,2,2-tetramethyldisilane ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4342-61-4
:
Formula:C₄H₁₂Cl₂Si₂
MDL Number:MFCD00078528
Boiling Pt:148°C
Flash Pt:37°C
EINECS:224-400-1
UN:2986
ADR:8,II
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together