
Didecyldimethylammonium bromide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2390-68-3
:
Formula:C₂₂H₄₈BrN
MDL Number:MFCD00012194
:406.54 g/mol
EINECS:219-234-1
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together