
Methyl-3-aminothiophene-2-carboxylate ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:22288-78-4
:
Formula:C₆H₇NO₂S
MDL Number:MFCD00009765
:157.19 g/mol
EINECS:244-895-8
:GHS07
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:64.0 to 68.0 deg-C
Specification Test Results
Frequently Bought Together