Order Entry
Netherlands
ContactUsLinkComponent
 
 
4-Methoxy-2-(trifluoromethyl)benzaldehyde ≥97.0% (by GC)
 :  TCI
 
undefined
 
4-Methoxy-2-(trifluoromethyl)benzaldehyde ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    38.0 to 42.0 deg-C

 

Frequently Bought Together