

4-Bromophenyl phenyl ether (BDE No. 3) ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:101-55-3
:
Formula:C₁₂H₉BrO
MDL Number:MFCD00000094
Boiling Pt:305°C
EINECS:202-952-4
UN:3082
ADR:9,III
:249.11 g/mol
:GHS07
:GHS09
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together