1,3-Bis(trimethylsilyl)urea ≥98.0% (by total nitrogen basis)
Catalog # TCIAB1103-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Solubility in hot Acetonitrile:almost transparency
Specification Test Results
About this item
:
:204.42 g/mol
EINECS:242-177-9
CAS Number:18297-63-7
Formula:C₇H₂₀N₂OSi₂
MDL Number:MFCD00008260