Glycylglycine (Gly-Gly)
Supplier: TCI
Certificates
About this item
- Melting point: 260 °C
- GHS symbol and signal word: GHS07 and warning
- EINECS: 209-127-8
- Merck index: 14,04503
- Solubility in water: Transparency
- Drying loss: Max. 0,2%
- Ignition residue (sulphate): Max. 0,05%
Product Categories: Bioscience, Buffer-related Reagents, Oligopeptides
Other Product Names / Synonyms: H-Gly-Gly-OH, Diglycine
Specifications
- CAS No.:556-50-3
- Storage conditions:Ambient
- Form:Crystal
- MDL No.:MFCD00008130
- Protein synonyms:Glycylglycine-Gly-Gly|H-Gly-Gly-OH|Diglycine
- Protein/peptide name:Glycylglycine (Gly-Gly)
- Purity:99,0 (titrimetric)
- Molecular weight:132,12 g/mol
- Molecular formula:C₄H₈N₂O₃
Specifications
Frequently Bought Together