Order Entry
Netherlands
ContactUsLinkComponent
Diuron (N-(3,4-dichlorophenyl)-N',N'-dimethylurea) ≥98.0% (by total nitrogen basis)
Irritant
Health
Environment
Diuron (N-(3,4-dichlorophenyl)-N',N'-dimethylurea) ≥98.0% (by total nitrogen basis)
Supplier:  TCI
Certificates
undefined
Irritant
Health
Environment
Diuron (N-(3,4-dichlorophenyl)-N',N'-dimethylurea) ≥98.0% (by total nitrogen basis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    157.0 to 160.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together