

4,4''-Diamino-p-terphenyl ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3365-85-3
:
Formula:C₁₈H₁₆N₂
MDL Number:MFCD00051532
:260.34 g/mol
Melting Pt:242°C
:GHS07
:GHS08
Specification Test Results
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together