Order Entry
Netherlands
ContactUsLinkComponent
3,3'-Iminobis(propylamine) ≥98.0% (by GC, titration analysis)
Corrosive
Toxic
3,3'-Iminobis(propylamine) ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Corrosive
Toxic
3,3'-Iminobis(propylamine) ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together