Calix[6]arene ≥95.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:96107-95-8
:
Formula:C₄₂H₃₆O₆
MDL Number:MFCD00143083
:636.74 g/mol
:GHS07
:GHS08
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 95.0 area%
Specification Test Results
Frequently Bought Together