(S)-(-)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:184346-45-0
:
Formula:C₁₈H₁₉NO₂
MDL Number:MFCD03093556
:281.35 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Optical purity (LC):min. 99.0 ee%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:252.0 to 256.0 deg-C
Specification Test Results
Frequently Bought Together