Sarcosine ethyl ester hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:52605-49-9
:
Formula:C₅H₁₁NO₂·HCl
MDL Number:MFCD00012506
:153.61 g/mol
EINECS:258-037-5
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:121.0 to 127.0 deg-C
Specification Test Results
Frequently Bought Together