Order Entry
Netherlands
ContactUsLinkComponent
 
 
1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene ≥95.0% (by GC)
 :  TCI
 
undefined
 
1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    White or Colorless to Yellow powder to lump to clear liquid
  • Purity (GC):
    min. 95.0 %

 

Frequently Bought Together