
trans,trans-Muconic acid (trans,trans-1,3-butadiene-1,4-dicarboxylic acid) ≥98.0% (by titrimetric analysis)
Catalog # TCIAM0473-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Solubility in hot Methanol:almost transparency
Specification Test Results
About this item
:
:142.11 g/mol
Merck Index:14,06299
EINECS:222-724-8
:GHS07
CAS Number:3588-17-8
Formula:C₆H₆O₄
MDL Number:MFCD00002702