Order Entry
Netherlands
ContactUsLinkComponent
Purpurin(1,2,4-trihydroxy-9,10-anthraquinone) ≥95.0% (by HPLC)
Irritant
Purpurin(1,2,4-trihydroxy-9,10-anthraquinone) ≥95.0% (by HPLC)
Catalog # TCIAP0605-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Purpurin(1,2,4-trihydroxy-9,10-anthraquinone) ≥95.0% (by HPLC)
Catalog # TCIAP0605-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Orange to Amber to Dark red powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Lambda max.:
    470.0 to 490.0 nm (MeOH)
  • Absorbance (E1%1cm):
    min. 300 (MeOH, 481.0 nm)

Specification Test Results

About this item

 :
Merck Index:14,07945
 :256.21 g/mol
EINECS:201-359-8
 :GHS07
CAS Number:81-54-9
Formula:C₁₄H₈O₅
MDL Number:MFCD00001203