
5-Hydroxy-1-tetralone ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:28315-93-7
:
Formula:C₁₀H₁₀O₂
MDL Number:MFCD00001693
:162.19 g/mol
EINECS:248-958-0
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:205.0 to 211.0 deg-C
Specification Test Results
Frequently Bought Together