Order Entry
Netherlands
ContactUsLinkComponent
 
6-Methoxy-2-tetralone ≥95.0% (by GC)
Catalog # TCIAM1317-1G
Leverancier:  TCI
CAS-nummer:  
undefined
6-Methoxy-2-tetralone ≥95.0% (by GC)
Catalog # TCIAM1317-1G
Leverancier:  TCI
CAS-nummer:  

Specificaties

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specificaties

Specificatie testresultaten

  • Appearance:
    White or Colorles to Yellow to Orange powder to lump to clear liquid
  • Purity (GC):
    min. 95.0 %

Specificatie testresultaten

Over dit artikel

 :
 :176.22 g/mol
Temperatuur bij opslag:Refrigerator
EINECS:219-592-9
CAS-nummer:2472-22-2
Formule:C₁₁H₁₂O₂
MDL-nummer:MFCD00001729