Order Entry
Netherlands
ContactUsLinkComponent
 
6-Methoxy-1-tetralone ≥99.0% (by GC)
 :  TCI
 
undefined
6-Methoxy-1-tetralone ≥99.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    77.0 to 81.0 deg-C

 

Frequently Bought Together