Order Entry
Netherlands
ContactUsLinkComponent
6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione ≥98.0% (by HPLC, titration analysis)
Irritant
6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together