Order Entry
Netherlands
ContactUsLinkComponent
 
 
Calcium thioglycolate trihydrate ≥94.0% (by titrimetric analysis)
 :  TCI
 
undefined
 
Calcium thioglycolate trihydrate ≥94.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Iodometric Titration):
    min. 94.0 % (calcd.on anh.substance)
  • Water:
    26.0 to 32.0 %

 

Frequently Bought Together