Order Entry
Netherlands
ContactUsLinkComponent
 
 
2-(Chloromethyl)-1,3-dioxolane ≥95.0% (by GC)
  TCIAC1321-25G
 :  TCI
 :  
undefined
 
2-(Chloromethyl)-1,3-dioxolane ≥95.0% (by GC)
  TCIAC1321-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 %

 

 

 :
 :122.55 g/mol
 :158°C
 :70°C
 :219-908-5
 :GHS07
 :2568-30-1
 :C₄H₇ClO₂
 :MFCD00043402