Order Entry
Netherlands
ContactUsLinkComponent
2-(Bromomethyl)-1,3-dioxolane ≥95.0% (by GC)
Irritant
2-(Bromomethyl)-1,3-dioxolane ≥95.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
2-(Bromomethyl)-1,3-dioxolane ≥95.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

Frequently Bought Together