4-Aminopiperidine ≥96.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13035-19-3
:
Formula:C₅H₁₂N₂
MDL Number:MFCD02179399
:100.16 g/mol
Flash Pt:48°C
UN:2925
ADR:4.1,II
:GHS02
:GHS05
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to lump
- Purity (GC):min. 96.0 %
- Purity (Nonaqueous Titration):min. 96.0 %
Specification Test Results
Frequently Bought Together