Order Entry
Netherlands
ContactUsLinkComponent
 
4-Aminobutyraldehyde diethyl acetal ≥98.0% (by GC, titration analysis)
 :  TCI
 
undefined
4-Aminobutyraldehyde diethyl acetal ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

Frequently Bought Together