
Methoxyhydroquinone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:824-46-4
:
Formula:C₇H₈O₃
MDL Number:MFCD00013971
:140.14 g/mol
Boiling Pt:180°C
EINECS:212-530-1
:GHS07
Specification Test Results
- Appearance:White to Light gray to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:88.0 to 91.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together