L(+)-Ornithine dihydrochloride ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:6211-16-1
:
Formula:C₅H₁₄Cl₂N₂O₂
MDL Number:MFCD00066571
:205.08 g/mol
Merck Index:14,06874
EINECS:228-277-5
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Specific rotation [a]20/D:+16.0 to +19.0 deg (C=5, H2O)
- Solubility in Water:almost transparency
Specification Test Results
Related Information
Frequently Bought Together