Order Entry
Netherlands
ContactUsLinkComponent
 
L(+)-Ornithine hydrochloride ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
L(+)-Ornithine hydrochloride ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +22.0 to +25.0 deg (C=4, 6mol/L HCl)
  • Solubility in Water:
    almost transparency

 

Frequently Bought Together