Trimethyl citrate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1587-20-8
:
Formula:C₉H₁₄O₇
MDL Number:MFCD00025889
:234.21 g/mol
Boiling Pt:176°C
EINECS:216-449-2
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:77.0 to 82.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together