
4,6-Dichloro-2-methylpyrimidine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1780-26-3
:
Formula:C₅H₄Cl₂N₂
MDL Number:MFCD00090472
:163.01 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:44.0 to 48.0 deg-C
Specification Test Results
Frequently Bought Together